Product Details

SNP ID
rs35110984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43650410 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGAGTGGAGCCTCTTGCCCCCTG[C/T]CACAGAGGAGATGGTGGCGCAGTCG
Phenotype
MIM: 612648
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAD2L1BP PubMed Links

Gene Details

Gene
MAD2L1BP
Gene Name
MAD2L1 binding protein
There are no transcripts associated with this gene.

Gene
RSPH9
Gene Name
radial spoke head 9 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193341.1 332 Missense Mutation GCC,GTC A88V NP_001180270.1
NM_152732.4 332 Missense Mutation GCC,GTC A88V NP_689945.2
XM_005248901.3 332 Missense Mutation GCC,GTC A88V XP_005248958.1
XM_006715014.1 332 Intron XP_006715077.1
XM_017010426.1 332 Missense Mutation GCC,GTC A88V XP_016865915.1

View Full Product Details