Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001099407.1 | 65 | Intron | NP_001092877.1 | ||
XM_005260742.3 | 65 | Intron | XP_005260799.1 | ||
XM_005260743.3 | 65 | Intron | XP_005260800.1 | ||
XM_006723584.2 | 65 | Intron | XP_006723647.1 | ||
XM_011529268.2 | 65 | Intron | XP_011527570.1 | ||
XM_011529271.2 | 65 | Intron | XP_011527573.1 | ||
XM_011529272.2 | 65 | Intron | XP_011527574.1 | ||
XM_011529275.2 | 65 | Intron | XP_011527577.1 | ||
XM_011529277.2 | 65 | Intron | XP_011527579.1 | ||
XM_017027909.1 | 65 | Intron | XP_016883398.1 | ||
XM_017027910.1 | 65 | Intron | XP_016883399.1 | ||
XM_017027911.1 | 65 | Intron | XP_016883400.1 | ||
XM_017027912.1 | 65 | Intron | XP_016883401.1 | ||
XM_017027913.1 | 65 | Intron | XP_016883402.1 | ||
XM_017027914.1 | 65 | Intron | XP_016883403.1 | ||
XM_017027915.1 | 65 | Intron | XP_016883404.1 | ||
XM_017027916.1 | 65 | Intron | XP_016883405.1 | ||
XM_017027917.1 | 65 | Intron | XP_016883406.1 | ||
XM_017027918.1 | 65 | Intron | XP_016883407.1 | ||
XM_017027919.1 | 65 | Intron | XP_016883408.1 | ||
XM_017027920.1 | 65 | Intron | XP_016883409.1 | ||
XM_017027921.1 | 65 | Intron | XP_016883410.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001282526.1 | 65 | UTR 5 | NP_001269455.1 | ||
NM_006466.3 | 65 | UTR 5 | NP_006457.2 | ||
XM_011529141.1 | 65 | UTR 5 | XP_011527443.1 | ||
XM_017027596.1 | 65 | UTR 5 | XP_016883085.1 |