Product Details

SNP ID
rs34013660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96877964 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGCCAGGTCCTTGAGCATGGTGC[A/G]GTGCATGGCCTGGGGGCTCTCCCGG
Phenotype
MIM: 604462
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM178B PubMed Links

Gene Details

Gene
FAM178B
Gene Name
family with sequence similarity 178 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122646.2 385 Missense Mutation CGC,TGC R645C NP_001116118.2
NM_001172667.1 385 Missense Mutation CGC,TGC R104C NP_001166138.1
NM_016490.4 385 Missense Mutation CGC,TGC R85C NP_057574.2
XM_011511294.2 385 Missense Mutation CGC,TGC R609C XP_011509596.1
XM_011511297.2 385 Intron XP_011509599.1
XM_017004263.1 385 Missense Mutation CGC,TGC R688C XP_016859752.1
XM_017004264.1 385 Missense Mutation CGC,TGC R675C XP_016859753.1
XM_017004265.1 385 Intron XP_016859754.1
XM_017004266.1 385 Missense Mutation CGC,TGC R111C XP_016859755.1
XM_017004267.1 385 Missense Mutation CGC,TGC R110C XP_016859756.1
XM_017004268.1 385 Missense Mutation CGC,TGC R85C XP_016859757.1
Gene
SEMA4C
Gene Name
semaphorin 4C
There are no transcripts associated with this gene.

View Full Product Details