Product Details

SNP ID
rs9261269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30062337 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTACACCTGTACCAACTGCAAGTG[A/G]GTATTCTTTCCCCTCCCTCTGCTCA
Phenotype
MIM: 606670 MIM: 607524 MIM: 607525 MIM: 615714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R11 PubMed Links

Gene Details

Gene
PPP1R11
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 11
There are no transcripts associated with this gene.

Gene
RNF39
Gene Name
ring finger protein 39
There are no transcripts associated with this gene.

Gene
ZNRD1
Gene Name
zinc ribbon domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278785.1 Intron NP_001265714.1
NM_001278786.1 Intron NP_001265715.1
NM_014596.5 Intron NP_055411.1
NM_170783.3 Intron NP_740753.1
Gene
ZNRD1ASP
Gene Name
zinc ribbon domain containing 1 antisense, pseudogene
There are no transcripts associated with this gene.

View Full Product Details