Product Details

SNP ID
rs79220020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74533079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCGAGACAGAGGGTTAAATGAA[C/T]CAGTGGGGGCAGGTCCCTCCAACCA
Phenotype
MIM: 602434 MIM: 606441 MIM: 607163
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AUP1 PubMed Links

Gene Details

Gene
AUP1
Gene Name
ancient ubiquitous protein 1
There are no transcripts associated with this gene.

Gene
DQX1
Gene Name
DEAQ-box RNA dependent ATPase 1
There are no transcripts associated with this gene.

Gene
HTRA2
Gene Name
HtrA serine peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321727.1 2455 UTR 3 NP_001308656.1
NM_001321728.1 2455 UTR 3 NP_001308657.1
NM_013247.4 2455 UTR 3 NP_037379.1
NM_145074.2 2455 UTR 3 NP_659540.1
Gene
LOXL3
Gene Name
lysyl oxidase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289164.1 2455 UTR 3 NP_001276093.1
NM_001289165.1 2455 UTR 3 NP_001276094.1
NM_032603.3 2455 UTR 3 NP_115992.1
XM_011533134.2 2455 Intron XP_011531436.1
XM_017005112.1 2455 Intron XP_016860601.1

View Full Product Details