Product Details

SNP ID
rs113931979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39063006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGTAGACTATATTTTATTACTGC[A/G]TGGCTTGTACTTTATAAAAAGCATA
Phenotype
MIM: 612167
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR48 PubMed Links

Gene Details

Gene
WDR48
Gene Name
WD repeat domain 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303402.1 Intron NP_001290331.1
NM_001303403.1 Intron NP_001290332.1
NM_020839.3 Intron NP_065890.1
XM_005265346.3 Intron XP_005265403.1
XM_011533971.1 Intron XP_011532273.1
XM_017006960.1 Intron XP_016862449.1

View Full Product Details