Product Details

SNP ID
rs34774892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:64686610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGACGGCCGTGTGCTCGGAGTT[A/G]GTATTGACATAGCCTTTGTAGAAAC
Phenotype
MIM: 605421
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS9 PubMed Links

Gene Details

Gene
ADAMTS9
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318781.1 506 Silent Mutation ACC,ACT T158T NP_001305710.1
NM_182920.1 506 Silent Mutation ACC,ACT T158T NP_891550.1
Gene
ADAMTS9-AS2
Gene Name
ADAMTS9 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details