Product Details

SNP ID
rs115701192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49741817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACACCAGCAGTACCCTTGAGTC[C/T]GGGGTTCCCCTCCCCTTGCCCCAAC
Phenotype
MIM: 608253
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSKS PubMed Links

Gene Details

Gene
TSKS
Gene Name
testis specific serine kinase substrate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021733.1 Intron NP_068379.1
XM_011527201.1 Intron XP_011525503.1
XM_011527202.1 Intron XP_011525504.1
XM_011527203.2 Intron XP_011525505.1

View Full Product Details