Product Details

SNP ID
rs34498516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124613081 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCACTGCTTCACCATAAGCAGGAC[C/G]GGCCTGGCCAGGACAAAATGAAATC
Phenotype
MIM: 608422 MIM: 610614
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PANX3 PubMed Links

Gene Details

Gene
PANX3
Gene Name
pannexin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052959.2 283 Missense Mutation CGG,GGG R95G NP_443191.1
Gene
TBRG1
Gene Name
transforming growth factor beta regulator 1
There are no transcripts associated with this gene.

View Full Product Details