Product Details

SNP ID
rs370083146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44926533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAGGAGGGAAGGTGGAAGGTTA[C/T]GGCCCTGTCTCTGGACTATGGGTGT
Phenotype
MIM: 614570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF18B PubMed Links

Gene Details

Gene
KIF18B
Gene Name
kinesin family member 18B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001264573.1 Intron NP_001251503.1
NM_001265577.1 Intron NP_001252506.1
XM_011524385.2 Intron XP_011522687.1
XM_011524386.2 Intron XP_011522688.1
XM_011524387.2 Intron XP_011522689.1
XM_011524388.2 Intron XP_011522690.1
XM_011524389.2 Intron XP_011522691.1
XM_011524390.2 Intron XP_011522692.1
XM_011524391.2 Intron XP_011522693.1
Gene
MIR6783
Gene Name
microRNA 6783
There are no transcripts associated with this gene.

View Full Product Details