Product Details

SNP ID
rs17714854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50375395 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCTGTCCTCCAGCACCAGAGTTA[C/T]TTTCACCACCTGTCCCAGAAATAGC
Phenotype
MIM: 610885 MIM: 614854 MIM: 611837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EME1 PubMed Links

Gene Details

Gene
EME1
Gene Name
essential meiotic structure-specific endonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166131.1 265 Missense Mutation CTT,TTT L63F NP_001159603.1
NM_152463.2 265 Missense Mutation CTT,TTT L63F NP_689676.2
XM_005257081.3 265 Missense Mutation CTT,TTT L63F XP_005257138.1
XM_011524395.2 265 UTR 5 XP_011522697.1
XM_017024236.1 265 Missense Mutation CTT,TTT L63F XP_016879725.1
Gene
LRRC59
Gene Name
leucine rich repeat containing 59
There are no transcripts associated with this gene.

Gene
MRPL27
Gene Name
mitochondrial ribosomal protein L27
There are no transcripts associated with this gene.

View Full Product Details