Product Details

SNP ID
rs34749181
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:115050530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTTTCGTATCGCACAGCAGAGG[A/G]CCATGCTGAATATCATGCCAAAGAT
Phenotype
MIM: 613133
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSPAN2 PubMed Links

Gene Details

Gene
TSPAN2
Gene Name
tetraspanin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308315.1 619 Missense Mutation GCC,GTC A184V NP_001295244.1
NM_001308316.1 619 Missense Mutation GCC,GTC A181V NP_001295245.1
NM_005725.5 619 Missense Mutation GCC,GTC A209V NP_005716.2
XM_016999996.1 619 Missense Mutation GCC,GTC A156V XP_016855485.1

View Full Product Details