Product Details

SNP ID
rs41267090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31880663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCAGAGCCCCTAGAGACCCCT[A/T]GAGTCTCACCCTAGCTCCTCCCCAG
Phenotype
MIM: 604599 MIM: 606107
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EHMT2 PubMed Links

Gene Details

Gene
EHMT2
Gene Name
euchromatic histone lysine methyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289413.1 Intron NP_001276342.1
NM_001318833.1 Intron NP_001305762.1
NM_006709.4 Intron NP_006700.3
NM_025256.6 Intron NP_079532.5
XM_005248824.3 Intron XP_005248881.1
XM_006714974.1 Intron XP_006715037.1
XM_006714975.1 Intron XP_006715038.1
XM_006714976.1 Intron XP_006715039.1
XM_017010202.1 Intron XP_016865691.1
Gene
SLC44A4
Gene Name
solute carrier family 44 member 4
There are no transcripts associated with this gene.

View Full Product Details