Product Details
- SNP ID
-
rs147304638
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:56015138 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTCCTTTTCCACCAGATTCAGCT[G/A]TTGGCTGCGTCTCATCAAAACCTGC
- Phenotype
-
MIM: 172480
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PSPH
PubMed Links
Gene Details
- Gene
- PSPH
- Gene Name
- phosphoserine phosphatase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004577.3 |
1184 |
Missense Mutation |
ACA,ATA |
T152I |
NP_004568.2 |
XM_005271773.1 |
1184 |
Missense Mutation |
ACA,ATA |
T152I |
XP_005271830.1 |
XM_005271774.1 |
1184 |
Missense Mutation |
ACA,ATA |
T152I |
XP_005271831.1 |
XM_005271775.1 |
1184 |
Missense Mutation |
ACA,ATA |
T152I |
XP_005271832.1 |
XM_005271776.1 |
1184 |
Missense Mutation |
ACA,ATA |
T152I |
XP_005271833.1 |
XM_006715760.1 |
1184 |
Missense Mutation |
ACA,ATA |
T152I |
XP_006715823.1 |
XM_006715761.1 |
1184 |
Missense Mutation |
ACA,ATA |
T107I |
XP_006715824.1 |
XM_011515461.1 |
1184 |
Missense Mutation |
ACA,ATA |
T152I |
XP_011513763.1 |
XM_017012466.1 |
1184 |
Missense Mutation |
ACA,ATA |
T152I |
XP_016867955.1 |
XM_017012467.1 |
1184 |
Missense Mutation |
ACA,ATA |
T152I |
XP_016867956.1 |
XM_017012468.1 |
1184 |
Missense Mutation |
ACA,ATA |
T107I |
XP_016867957.1 |
XM_017012469.1 |
1184 |
Missense Mutation |
ACA,ATA |
T107I |
XP_016867958.1 |
View Full Product Details