Product Details

SNP ID
rs143267328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2168045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCCACATCTGCTGAGGGAGCC[C/T]CCACTGAGACGCCAGCCCTCTTGCC
Phenotype
MIM: 612184 MIM: 606692
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASKIN1 PubMed Links

Gene Details

Gene
CASKIN1
Gene Name
CASK interacting protein 1
There are no transcripts associated with this gene.

Gene
TRAF7
Gene Name
TNF receptor associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032271.2 Intron NP_115647.2
XM_005255627.4 Intron XP_005255684.1
XM_011522700.1 Intron XP_011521002.1

View Full Product Details