Product Details

SNP ID
rs61742675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104695051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAATAATGTCATTGATCATATTAC[C/T]TGTGAAATTTTGGCCCTTCTAAAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR13D1 PubMed Links

Gene Details

Gene
OR13D1
Gene Name
olfactory receptor family 13 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004484.1 630 Silent Mutation ACC,ACT T210T NP_001004484.1

View Full Product Details