Product Details

SNP ID
rs61730237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:185511676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCAAGTAGACTCACTTGGTGATA[C/T]TTCTGAAATGTGGTGGGTTCACTGG
Phenotype
MIM: 607365
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIPH PubMed Links

Gene Details

Gene
LIPH
Gene Name
lipase H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139248.2 1355 Silent Mutation AAA,AAG K372K NP_640341.1
XM_006713529.3 1355 Silent Mutation AAA,AAG K342K XP_006713592.1
XM_011512530.2 1355 Silent Mutation AAA,AAG K329K XP_011510832.1
XM_011512531.2 1355 Silent Mutation AAA,AAG K329K XP_011510833.1
XM_017005852.1 1355 Silent Mutation AAA,AAG K338K XP_016861341.1

View Full Product Details