Product Details

SNP ID
rs61734996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49739863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCCCGTGGACCCCTCAAGTGG[C/G]AGGTTGCTGAGATGATCGTGGAGGG
Phenotype
MIM: 608253
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TSKS PubMed Links

Gene Details

Gene
TSKS
Gene Name
testis specific serine kinase substrate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021733.1 1703 Silent Mutation CTC,CTG L564L NP_068379.1
XM_011527201.1 1703 Silent Mutation CTC,CTG L589L XP_011525503.1
XM_011527202.1 1703 Missense Mutation CCC,GCC P579A XP_011525504.1
XM_011527203.2 1703 UTR 3 XP_011525505.1

View Full Product Details