Product Details

SNP ID
rs10417344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36686697 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGGGACACTTACAACTCGAAAAG[C/G]CTCGTTCCGAAAACAGGTCTGTCCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC01534 PubMed Links

Gene Details

Gene
LINC01534
Gene Name
long intergenic non-protein coding RNA 1534
There are no transcripts associated with this gene.

Gene
ZNF567
Gene Name
zinc finger protein 567
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300979.1 Intron NP_001287908.1
NM_001322911.1 Intron NP_001309840.1
NM_001322912.1 Intron NP_001309841.1
NM_001322913.1 Intron NP_001309842.1
NM_001322914.1 Intron NP_001309843.1
NM_001322915.1 Intron NP_001309844.1
NM_001322916.1 Intron NP_001309845.1
NM_001322917.1 Intron NP_001309846.1
NM_001322918.1 Intron NP_001309847.1
NM_001322919.1 Intron NP_001309848.1
NM_001322920.1 Intron NP_001309849.1
NM_152603.4 Intron NP_689816.2
XM_011526584.2 Intron XP_011524886.1
XM_011526585.2 Intron XP_011524887.1
XM_017026416.1 Intron XP_016881905.1
XM_017026417.1 Intron XP_016881906.1
XM_017026418.1 Intron XP_016881907.1
XM_017026419.1 Intron XP_016881908.1
XM_017026420.1 Intron XP_016881909.1
XM_017026421.1 Intron XP_016881910.1

View Full Product Details