Product Details
- SNP ID
-
rs61753623
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:110875559 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAGAGGATCTGCCTAAAGGTGGC[A/G]CTCATTTCTTTGTCGCGGTAGGAGT
- Phenotype
-
MIM: 602282
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LPAR1
PubMed Links
Gene Details
- Gene
- LPAR1
- Gene Name
- lysophosphatidic acid receptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001401.3 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
NP_001392.2 |
NM_057159.2 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
NP_476500.1 |
XM_005251781.3 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_005251838.1 |
XM_005251782.3 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_005251839.1 |
XM_017014383.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869872.1 |
XM_017014384.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869873.1 |
XM_017014385.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869874.1 |
XM_017014386.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869875.1 |
XM_017014387.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869876.1 |
XM_017014388.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869877.1 |
XM_017014389.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869878.1 |
XM_017014390.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869879.1 |
XM_017014391.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869880.1 |
XM_017014392.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869881.1 |
XM_017014393.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869882.1 |
XM_017014394.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869883.1 |
XM_017014395.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869884.1 |
XM_017014396.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869885.1 |
XM_017014397.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869886.1 |
XM_017014398.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869887.1 |
XM_017014399.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869888.1 |
XM_017014400.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869889.1 |
XM_017014401.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869890.1 |
XM_017014402.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869891.1 |
XM_017014403.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869892.1 |
XM_017014404.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869893.1 |
XM_017014405.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869894.1 |
XM_017014406.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869895.1 |
XM_017014407.1 |
1341 |
Silent Mutation |
AGC,AGT |
S319S |
XP_016869896.1 |
View Full Product Details