Product Details

SNP ID
rs6681483
Assay Type
Functionally tested
NCBI dbSNP Submissions
55
Location
Chr.1:247840994 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACATTTCCTTCTCATGACCTTTCT[A/G]ACAGCTTCTTTGAAGTCTTTGTTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR11L1 PubMed Links

Gene Details

Gene
OR11L1
Gene Name
olfactory receptor family 11 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001959.1 903 Silent Mutation GTC,GTT V301V NP_001001959.1

View Full Product Details