Product Details

SNP ID
rs11539159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109903047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGATGGCGCTCCCGACGCTGCC[A/G]TCCTACTGGTGCAGCCAGCAGCGCC
Phenotype
MIM: 612654
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCHP PubMed Links

Gene Details

Gene
TCHP
Gene Name
trichoplein keratin filament binding
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143852.1 198 Silent Mutation CCA,CCG P7P NP_001137324.1
NM_032300.4 198 Silent Mutation CCA,CCG P7P NP_115676.1
XM_011538836.2 198 Silent Mutation CCA,CCG P7P XP_011537138.1
XM_011538837.2 198 Silent Mutation CCA,CCG P7P XP_011537139.1
XM_017020024.1 198 Silent Mutation CCA,CCG P7P XP_016875513.1
XM_017020025.1 198 UTR 5 XP_016875514.1

View Full Product Details