Product Details
- SNP ID
-
rs75657243
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:108755720 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACGTTTGAGTTAGATTTCTTGGTT[G/T]GGACCCTGGTTTCATTACTGTCATG
- Phenotype
-
MIM: 605645
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RETNLB
PubMed Links
Gene Details
- Gene
- RETNLB
- Gene Name
- resistin like beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032579.2 |
492 |
UTR 3 |
|
|
NP_115968.1 |
View Full Product Details