Product Details

SNP ID
rs57228841
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.21:14273869 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TCCACCAGCACCCCGCCCCAGGCGC[A/T]CTGGCTAGGACTGGAAGGCAAGGAC
Phenotype
MIM: 608835
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCC13 PubMed Links

Gene Details

Gene
ABCC13
Gene Name
ATP binding cassette subfamily C member 13 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details