Product Details

SNP ID
rs7018553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104504543 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTCAGGGAGAAACACTATTTCAT[C/T]CTCAGGGTGCGCCACTCAGATGTAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR13F1 PubMed Links

Gene Details

Gene
OR13F1
Gene Name
olfactory receptor family 13 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004485.1 281 Missense Mutation TCC,TTC S94F NP_001004485.1

View Full Product Details