Product Details

SNP ID
rs80123117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14053845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGGGGCCTTGGCCCTCAGCAGC[A/G]GTTTGGGGCTTCACTCCTTCCTCCT
Phenotype
MIM: 605350
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL27RA PubMed Links

Gene Details

Gene
IL27RA
Gene Name
interleukin 27 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004843.3 1782 Intron NP_004834.1
Gene
PALM3
Gene Name
paralemmin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145028.1 1782 Silent Mutation ACC,ACT T594T NP_001138500.1
XM_005259891.4 1782 Silent Mutation ACC,ACT T609T XP_005259948.1
XM_011527965.2 1782 Silent Mutation ACC,ACT T582T XP_011526267.1

View Full Product Details