Product Details
- SNP ID
-
rs17302903
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:55509161 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCGCCCGCGTAGATCGCTTCGGCCG[C/G]GTTCTACGCCCGGCTCAACTATGAG
- Phenotype
-
MIM: 612040
MIM: 120360
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LPCAT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150333127] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LPCAT2
- Gene Name
- lysophosphatidylcholine acyltransferase 2
- Gene
- MMP2
- Gene Name
- matrix metallopeptidase 2
There are no transcripts associated with this gene.
View Full Product Details