Product Details

SNP ID
rs73362716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142199769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTAGCCCCTTCATCAGAGGTAA[A/G]AGGGTATCCCCATTCCCACCCGCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00051 PubMed Links

Gene Details

Gene
LINC00051
Gene Name
long intergenic non-protein coding RNA 51
There are no transcripts associated with this gene.

View Full Product Details