Product Details
- SNP ID
-
hCV25989459
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:185367349 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTCATTTTTAAGAATAGTAAAGT[A/C]CAGTGAATTTCAAGAAAATTTGTTA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LRP2BP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77624707] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRP2BP
- Gene Name
- LRP2 binding protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018409.3 |
|
Intron |
|
|
NP_060879.2 |
- Gene
- SNX25
- Gene Name
- sorting nexin 25
View Full Product Details