Product Details

SNP ID
rs34540158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35194257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCTGGGTGGAGCGGAGGGCGG[C/T]GATGGGTGTGTGGAGTCAGGCTGGT
Phenotype
MIM: 611220
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC35G3 PubMed Links

Gene Details

Gene
SLC35G3
Gene Name
solute carrier family 35 member G3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152462.2 137 Silent Mutation NP_689675.1
Gene
UNC45B
Gene Name
unc-45 myosin chaperone B
There are no transcripts associated with this gene.

View Full Product Details