Product Details

SNP ID
rs62441827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149963992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGCAGCTTTATAAACAGCCGT[C/G]GTGTGAGCCTCGAAGGGAACCATCA
Phenotype
MIM: 605697
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ULBP1 PubMed Links

Gene Details

Gene
ULBP1
Gene Name
UL16 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317089.1 19 Intron NP_001304018.1
NM_025218.3 19 Intron NP_079494.1
XM_017011322.1 19 UTR 5 XP_016866811.1

View Full Product Details