Product Details

SNP ID
rs7527057
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:74706317 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCATCATAAGAGAAGAATCATTTT[A/T]CCAGTAGCCCCACTACCATGAATGA
Phenotype
MIM: 123691
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CRYZ PubMed Links

Gene Details

Gene
CRYZ
Gene Name
crystallin zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130042.1 958 Silent Mutation GGA,GGT G323G NP_001123514.1
NM_001130043.1 958 Silent Mutation GGA,GGT G289G NP_001123515.1
NM_001134759.1 958 Silent Mutation GGA,GGT G186G NP_001128231.1
NM_001889.3 958 Silent Mutation GGA,GGT G323G NP_001880.2
XM_005270491.4 958 Silent Mutation GGA,GGT G186G XP_005270548.1
XM_011540747.1 958 Silent Mutation GGA,GGT G323G XP_011539049.1
XM_017000367.1 958 Silent Mutation GGA,GGT G289G XP_016855856.1
XM_017000368.1 958 Silent Mutation GGA,GGT G186G XP_016855857.1

View Full Product Details