Product Details

SNP ID
rs369034392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101327858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAGAGTGACTAAAGTAGAAATCT[A/C]TTTATCAGTATGCAACATTTTCAGA
Phenotype
MIM: 607056 MIM: 612846
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IMPG2 PubMed Links

Gene Details

Gene
IMPG2
Gene Name
interphotoreceptor matrix proteoglycan 2
There are no transcripts associated with this gene.

Gene
SENP7
Gene Name
SUMO1/sentrin specific peptidase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077203.2 Intron NP_001070671.1
NM_001282801.1 Intron NP_001269730.1
NM_001282802.1 Intron NP_001269731.1
NM_001282803.1 Intron NP_001269732.1
NM_001282804.1 Intron NP_001269733.1
NM_020654.4 Intron NP_065705.3
XM_005247662.4 Intron XP_005247719.1
XM_011513038.2 Intron XP_011511340.1
XM_011513039.2 Intron XP_011511341.1
XM_011513040.2 Intron XP_011511342.1
XM_011513041.2 Intron XP_011511343.1
XM_011513042.2 Intron XP_011511344.1
XM_011513045.2 Intron XP_011511347.1
XM_017006924.1 Intron XP_016862413.1
XM_017006925.1 Intron XP_016862414.1
XM_017006926.1 Intron XP_016862415.1
XM_017006927.1 Intron XP_016862416.1
XM_017006928.1 Intron XP_016862417.1
XM_017006929.1 Intron XP_016862418.1

View Full Product Details