Product Details

SNP ID
rs111413504
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:161119106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTGCTTCCCTGTGCTATGAAAT[A/C]TGAGAATCCTGCCTATGCTGTTCAC
Phenotype
MIM: 606841 MIM: 604618 MIM: 613466
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DEDD PubMed Links

Gene Details

Gene
DEDD
Gene Name
death effector domain containing
There are no transcripts associated with this gene.

Gene
NIT1
Gene Name
nitrilase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185092.1 Intron NP_001172021.1
NM_001185093.1 Intron NP_001172022.1
NM_001185094.1 Intron NP_001172023.1
NM_005600.2 Intron NP_005591.1
XM_005245214.2 Intron XP_005245271.1
XM_005245215.4 Intron XP_005245272.1
XM_005245216.3 Intron XP_005245273.1
XM_006711341.3 Intron XP_006711404.1
XM_017001371.1 Intron XP_016856860.1
Gene
PFDN2
Gene Name
prefoldin subunit 2
There are no transcripts associated with this gene.

View Full Product Details