Product Details

SNP ID
rs61739598
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:108718880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTATATATGGCAATTGCATTCTC[C/T]GTGTCCATTATGCGAGCCAATGGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FNDC7 PubMed Links

Gene Details

Gene
FNDC7
Gene Name
fibronectin type III domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144937.1 429 Silent Mutation TCC,TCT S143S NP_001138409.1

View Full Product Details