Product Details

SNP ID
rs61739615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7855533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTCACCGGATTCCTGCTCCTCG[G/T]GCCGCTGCTCGTGCTTGCTCTGGCC
Phenotype
MIM: 611577
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYB5D1 PubMed Links

Gene Details

Gene
CYB5D1
Gene Name
cytochrome b5 domain containing 1
There are no transcripts associated with this gene.

Gene
KDM6B
Gene Name
lysine demethylase 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080424.1 308 Intron NP_001073893.1
XM_005256549.3 308 Intron XP_005256606.1
XM_005256550.4 308 Intron XP_005256607.1
XM_005256551.3 308 Intron XP_005256608.1
XM_005256552.4 308 Intron XP_005256609.1
XM_006721483.3 308 Intron XP_006721546.1
XM_011523750.2 308 Intron XP_011522052.1
XM_011523752.2 308 Intron XP_011522054.1
XM_017024381.1 308 Intron XP_016879870.1
Gene
NAA38
Gene Name
N(alpha)-acetyltransferase 38, NatC auxiliary subunit
There are no transcripts associated with this gene.

Gene
TMEM88
Gene Name
transmembrane protein 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319941.1 308 Intron NP_001306870.1
NM_203411.1 308 Missense Mutation GGG,GTG G100V NP_981956.1
XM_005256856.3 308 Missense Mutation GGG,GTG G114V XP_005256913.1

View Full Product Details