Product Details

SNP ID
rs116796737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61337880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTACACCACAGCAGTGTGGCTG[C/T]GCAGGATGGGGGTATCTGTACTGAC
Phenotype
MIM: 600045 MIM: 615844
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDB1 PubMed Links

Gene Details

Gene
DDB1
Gene Name
damage specific DNA binding protein 1
There are no transcripts associated with this gene.

Gene
TKFC
Gene Name
triokinase and FMN cyclase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015533.3 Intron NP_056348.2
XM_011544912.2 Intron XP_011543214.1
XM_017017517.1 Intron XP_016873006.1
XM_017017518.1 Intron XP_016873007.1
XM_017017519.1 Intron XP_016873008.1
XM_017017520.1 Intron XP_016873009.1
XM_017017521.1 Intron XP_016873010.1
XM_017017522.1 Intron XP_016873011.1
XM_017017523.1 Intron XP_016873012.1
XM_017017524.1 Intron XP_016873013.1

View Full Product Details