Product Details

SNP ID
rs60681475
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.16:48170219 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
GCAGCTTTTTGGGGAACTTTGAGAT[C/T]TGCGATATGGGAAGAAGAGACAACA
Phenotype
MIM: 607040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCC11 PubMed Links

Gene Details

Gene
ABCC11
Gene Name
ATP binding cassette subfamily C member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032583.3 Intron NP_115972.2
NM_033151.3 Intron NP_149163.2
NM_145186.2 Intron NP_660187.1
XM_011523397.1 Intron XP_011521699.1
XM_011523398.2 Intron XP_011521700.1
XM_017023795.1 Intron XP_016879284.1
XM_017023796.1 Intron XP_016879285.1
XM_017023797.1 Intron XP_016879286.1
XM_017023798.1 Intron XP_016879287.1
XM_017023799.1 Intron XP_016879288.1
XM_017023800.1 Intron XP_016879289.1
XM_017023801.1 Intron XP_016879290.1
XM_017023802.1 Intron XP_016879291.1
XM_017023803.1 Intron XP_016879292.1

View Full Product Details