Product Details

SNP ID
rs73265622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64468299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATGAATATGCCTGGAAGAAATT[G/T]TACCTAGTGTGACATTTTTGCTCAT
Phenotype
MIM: 604688 MIM: 172460 MIM: 194541
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AKAP5 PubMed Links

Gene Details

Gene
AKAP5
Gene Name
A-kinase anchoring protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004857.3 283 UTR 5 NP_004848.3
Gene
MTHFD1
Gene Name
methylenetetrahydrofolate dehydrogenase, cyclohydrolase and formyltetrahydrofolate synthetase 1
There are no transcripts associated with this gene.

Gene
ZBTB25
Gene Name
zinc finger and BTB domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304507.1 283 Intron NP_001291436.1
NM_001304508.1 283 Intron NP_001291437.1
NM_006977.3 283 Intron NP_008908.2
XM_005268051.3 283 Intron XP_005268108.1
XM_006720250.3 283 Intron XP_006720313.1
XM_006720251.3 283 Intron XP_006720314.1
XM_006720252.3 283 Intron XP_006720315.1
XM_011537139.1 283 Intron XP_011535441.1
XM_011537141.2 283 Intron XP_011535443.1
XM_011537143.2 283 Intron XP_011535445.1
XM_011537146.2 283 Intron XP_011535448.1
XM_017021630.1 283 Intron XP_016877119.1
XM_017021631.1 283 Intron XP_016877120.1
XM_017021632.1 283 Intron XP_016877121.1
XM_017021633.1 283 Intron XP_016877122.1
XM_017021634.1 283 Intron XP_016877123.1
XM_017021635.1 283 Intron XP_016877124.1
XM_017021636.1 283 Intron XP_016877125.1
XM_017021637.1 283 Intron XP_016877126.1
XM_017021638.1 283 Intron XP_016877127.1

View Full Product Details