Product Details

SNP ID
rs115829004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15283314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTAAATGGTTTCTACTTGCCCCT[G/T]GGTTAATGAGAACATTAAACTAACC
Phenotype
MIM: 300626
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASB11 PubMed Links

Gene Details

Gene
ASB11
Gene Name
ankyrin repeat and SOCS box containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012428.2 1131 UTR 3 NP_001012428.1
NM_001201583.1 1131 UTR 3 NP_001188512.1
NM_080873.2 1131 UTR 3 NP_543149.1

View Full Product Details