Product Details

SNP ID
rs113881345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10489863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTCTGGCTCATACCTGCAAGGG[C/T]GTAAGAAAAATGGGGACAATGGCCA
Phenotype
MIM: 606016
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KEAP1 PubMed Links

Gene Details

Gene
KEAP1
Gene Name
kelch like ECH associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012289.3 Intron NP_036421.2
NM_203500.1 Intron NP_987096.1
XM_005260173.1 Intron XP_005260230.1
XM_005260174.1 Intron XP_005260231.1
XM_011528452.1 Intron XP_011526754.1

View Full Product Details