Product Details

SNP ID
rs56076213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110135108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAAGATTCAGCCAAGCATGAGT[A/G]TACATCTGAGGATGGACTCAAATAA
Phenotype
MIM: 605489
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT81 PubMed Links

Gene Details

Gene
IFT81
Gene Name
intraflagellar transport 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143779.1 Intron NP_001137251.1
NM_014055.3 Intron NP_054774.2
NM_031473.3 Intron NP_113661.2
XM_011538232.2 Intron XP_011536534.1
XM_017019217.1 Intron XP_016874706.1
XM_017019218.1 Intron XP_016874707.1

View Full Product Details