Product Details

SNP ID
rs7501060
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20538778 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGCAAATAAGGAAACTGAGGAAC[A/G]CAGAGGTTATCGAGTAAGTGGTGGA
Phenotype
MIM: 614359
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ACSM2B PubMed Links
Additional Information
For this assay, SNP(s) [rs114614696] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ACSM2B
Gene Name
acyl-CoA synthetase medium-chain family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105069.1 Intron NP_001098539.1
NM_182617.3 Intron NP_872423.3
XM_005255304.3 Intron XP_005255361.1
XM_011545824.2 Intron XP_011544126.1
XM_017023205.1 Intron XP_016878694.1

View Full Product Details