Product Details
- SNP ID
-
rs28623434
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
35
- Location
-
Chr.1:1618213 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCACTTCCCATAGCAGCCTCTGG[G/C]CCAGAGCTCACCCGCTGTGGGCAGG
- Phenotype
-
MIM: 611141
- Polymorphism
- G/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142389064] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIB2
- Gene Name
- mindbomb E3 ubiquitin protein ligase 2
View Full Product Details