Product Details
- SNP ID
-
rs12948507
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:82049081 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCTCCAGCCACGTTCACCACGTT[A/C]TGTGGGTTCTCAGGACCCCCATGGC
- Phenotype
-
MIM: 601934
MIM: 602578
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DUS1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74659471] are located under a probe and SNP(s) [rs78793795] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DUS1L
- Gene Name
- dihydrouridine synthase 1 like
There are no transcripts associated with this gene.
- Gene
- GPS1
- Gene Name
- G protein pathway suppressor 1
There are no transcripts associated with this gene.
- Gene
- RFNG
- Gene Name
- RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
View Full Product Details