Product Details

SNP ID
rs4604109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56654323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTAATTATGCTGCTTTATAGGTA[A/T]GATGACTGATGGCAACTCGCCAGGC
Phenotype
MIM: 607275
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HOPX PubMed Links

Gene Details

Gene
HOPX
Gene Name
HOP homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145459.1 Intron NP_001138931.1
NM_001145460.1 Intron NP_001138932.1
NM_032495.5 Intron NP_115884.4
NM_139211.4 Intron NP_631957.1
NM_139212.3 Intron NP_631958.1
XM_006714052.2 Intron XP_006714115.1
XM_017008728.1 Intron XP_016864217.1
XM_017008729.1 Intron XP_016864218.1
XM_017008730.1 Intron XP_016864219.1
XM_017008731.1 Intron XP_016864220.1
XM_017008732.1 Intron XP_016864221.1
XM_017008733.1 Intron XP_016864222.1
XM_017008734.1 Intron XP_016864223.1

View Full Product Details