Product Details

SNP ID
rs4859887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:74937902 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATAATGTGCCAACTCAGATTTAG[C/T]GCAAGAGTCCAAAAAATGCTCAGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARM1 PubMed Links

Gene Details

Gene
PARM1
Gene Name
prostate androgen-regulated mucin-like protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015393.3 Intron NP_056208.2
XM_011531833.1 Intron XP_011530135.1

View Full Product Details