Product Details

SNP ID
rs4419353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113536090 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGAAACCTAGGCCATACTGTTCC[A/G]ACAATTTACCACCAGGGGGATTTCA
Phenotype
MIM: 606816
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIDT1 PubMed Links

Gene Details

Gene
SIDT1
Gene Name
SID1 transmembrane family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308350.1 Intron NP_001295279.1
NM_001322294.1 Intron NP_001309223.1
NM_001322295.1 Intron NP_001309224.1
NM_001322296.1 Intron NP_001309225.1
NM_001322297.1 Intron NP_001309226.1
NM_001322298.1 Intron NP_001309227.1
NM_001322299.1 Intron NP_001309228.1
NM_001322300.1 Intron NP_001309229.1
NM_017699.2 Intron NP_060169.2
XM_011512939.2 Intron XP_011511241.1
XM_011512941.2 Intron XP_011511243.1
XM_017006664.1 Intron XP_016862153.1
XM_017006665.1 Intron XP_016862154.1
XM_017006666.1 Intron XP_016862155.1
XM_017006667.1 Intron XP_016862156.1
XM_017006668.1 Intron XP_016862157.1

View Full Product Details