Product Details

SNP ID
rs9895984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81170231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTGTGTTGGTATGTGTGCATGT[A/G]TGTGTACACAGGTGTGTGTGCATGT
Phenotype
MIM: 605276
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AATK PubMed Links

Gene Details

Gene
AATK
Gene Name
apoptosis associated tyrosine kinase
There are no transcripts associated with this gene.

Gene
AATK-AS1
Gene Name
AATK antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details