Product Details

SNP ID
rs4236749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:130052907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGACACTGGTGTGGCTTCAGTAC[C/T]GAGGCTGCTAAAGCTGCCAGTCACA
Phenotype
MIM: 605953
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASAP1 PubMed Links

Gene Details

Gene
ASAP1
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001247996.1 5280 UTR 3 NP_001234925.1
NM_018482.3 5280 UTR 3 NP_060952.2
XM_005250925.1 5280 UTR 3 XP_005250982.1
XM_006716563.3 5280 UTR 3 XP_006716626.1
XM_006716564.1 5280 UTR 3 XP_006716627.1
XM_006716565.3 5280 UTR 3 XP_006716628.1
XM_006716566.1 5280 UTR 3 XP_006716629.1
XM_011517052.2 5280 UTR 3 XP_011515354.1
XM_011517053.1 5280 UTR 3 XP_011515355.1
XM_017013467.1 5280 UTR 3 XP_016868956.1
XM_017013468.1 5280 UTR 3 XP_016868957.1
XM_017013469.1 5280 UTR 3 XP_016868958.1

View Full Product Details